Learn More
Indigo Instruments PEROXIDE TEST STRIPS 100/VIAL DFS Item
Supplier:  Indigo Instruments 33815P10PC
The Hydrogen Peroxide test trips are ideal for the evaluation of hydrogen peroxide concentrations up to 10%. The strips change its color to black and the speed of the color change indicates hydrogen peroxide's concentration. This product is packaged in a vial and is sold in a pack of 50.
Catalog No. NC9966580
Only null left
Explore available promotions