Learn More
Haztech Systems Inc Hazcat Peroxide Test Strips
Catalog No. NC9253631
Encompass
Hazcat Peroxide Test Strips. Test strips for quick and accurate detection of peroxide compounds. Can detect ether or THF solvents with high peroxide content, MEKP, Pirahna acid, and hydrogen peroxide, etc.. Amber glass bottle contains 10 Test Strips.
Catalog No. NC9253631 Supplier Haztech Systems Inc Supplier No. HW4047
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Hazcat Peroxide Test Strips. Test strips for quick and accurate detection of peroxide compounds. Can detect ether or THF solvents with high peroxide content, MEKP, Pirahna acid, and hydrogen peroxide, etc.. Amber glass bottle contains 10 Test Strips.