Learn More
GA International Permanent Deep-Freeze Labels for Thermal-Transfer Barcode Printers 1"" x 0.75"" / 25.4mm x 19.1mm Rectangle White From -80C to +110C (-112F to +230F)
Catalog No. 5000571642
Encompass
Deep-freeze, permanent barcode labels for small diameter vials, microtubes, falcon tubes, cell culture plates, microplates, as well as polypropylene and cardboard freezer boxes. The freezer-grade labels are waterproof and tearproof, can adhere to moist and wet surfaces, and will permanently adhere to containers in (-20C, -40C, -80C) freezers and dry ice without the need for extra lamination. Available in multiple configurations and colors, they are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, surface cleaning wipes, and other chemicals.
Catalog No. 5000571642 Supplier GA International Supplier No. FJT4C11WH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Deep-freeze, permanent barcode labels for small diameter vials, microtubes, falcon tubes, cell culture plates, microplates, as well as polypropylene and cardboard freezer boxes. The freezer-grade labels are waterproof and tearproof, can adhere to moist and wet surfaces, and will permanently adhere to containers in (-20C, -40C, -80C) freezers and dry ice without the need for extra lamination. Available in multiple configurations and colors, they are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, surface cleaning wipes, and other chemicals.