Learn More
GA International Permanent Deep-Freeze Labels for Thermal-Transfer Barcode Printers 1.4"" x 1"" / 35.6mm x 25.4mm Rectangle Lemon From -80C to +110C (-112F to +230F)
Catalog No. 5000571478
Encompass
Deep-freeze, permanent barcode labels for small diameter vials, microtubes, falcon tubes, cell culture plates, microplates, as well as polypropylene and cardboard freezer boxes. The freezer-grade labels are waterproof and tearproof, can adhere to moist and wet surfaces, and will permanently adhere to containers in (-20C, -40C, -80C) freezers and dry ice without the need for extra lamination. Available in multiple configurations and colors, they are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, surface cleaning wipes, and other chemicals.
Catalog No. 5000571478 Supplier GA International Supplier No. FJT243C34LE*
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Deep-freeze, permanent barcode labels for small diameter vials, microtubes, falcon tubes, cell culture plates, microplates, as well as polypropylene and cardboard freezer boxes. The freezer-grade labels are waterproof and tearproof, can adhere to moist and wet surfaces, and will permanently adhere to containers in (-20C, -40C, -80C) freezers and dry ice without the need for extra lamination. Available in multiple configurations and colors, they are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, surface cleaning wipes, and other chemicals.