Learn More
GA International Permanent Autoclave-Resistant Labels for Laser Printers 8.5"" x 11"" / 215.9mm x 279.4mm Rectangle White From -40C to +150C (-40F to +302F)
Catalog No. 5000571391
Encompass
Permanent laser printable labels, ideal for glassware, surgical instruments, metal and plastic surfaces, trays, blister packs, and other items intended for use in steam autoclaves, ovens, and gamma irradiators. Our autoclave laser labels are durable, inert, weather & water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121C/250F, 16.5 psi for 20 minutes). Compatible with all major brands of desktop laser printers.
Catalog No. 5000571391 Supplier GA International Supplier No. AKA28WH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Permanent laser printable labels, ideal for glassware, surgical instruments, metal and plastic surfaces, trays, blister packs, and other items intended for use in steam autoclaves, ovens, and gamma irradiators. Our autoclave laser labels are durable, inert, weather & water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121C/250F, 16.5 psi for 20 minutes). Compatible with all major brands of desktop laser printers.