Learn More
Contec™ PeridoxRTU™ Sporicidal Disinfectant
PeridoxRTU ™ is a simple one-step cleaner, disinfectant and sporicide.
Supplier:  Contec™ CR85365
Catalog No. 296366885A
Only null left
Explore available promotions
Description
Description
  • Broad spectrum disinfectant effective against bacterial spores, bacteria, viruses, TB and fungi (mold).
  • Excellent material compatibility that will not damage most surfaces in cleanroom environments, including stainless steel, acrylic, glass, aluminum and chrome.
  • Does not contain alcohol, bleach, quat or silver.
  • Daily cleaning and disinfecting tasks, such as wiping surfaces and mopping floors.
  • Effective component to monthly cleaning protocols.
  • PeridoxRTU is ideal for use in pharmaceutical, biopharmaceutical and medical device facilities.
  • Sterile PeridoxRTU is ideal for pharmaceutical and life science cleanroom environments, including sterile core areas.
  • Wear appropriate gloves and eye protection. Additional PPE may be required based on engineering controls/environment, method of application, and use protocols.
Order Info
Note: This part (19-033-460) is available in Fisher Canada only.
Product Suggestions
Product Suggestions