missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PeridoxRTU™ Sporicidal Disinfectant

Catalog No. p-9023001
Change view
Click to view available options
Volume (English):
32 oz.
1 gal.
Sterility:
Sterile
Packaging:
Double bagged
Double bagged with push/pull cap and foaming trigger
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Sterility Packaging
296366887AA 32 oz. Sterile Double bagged with push/pull cap and foaming trigger
19033463 1 gal. Sterile Double bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296366887AA Supplier Contec™ Supplier No. CR85365IR

PeridoxRTU ™ is a simple one-step cleaner, disinfectant and sporicide.

  • Broad spectrum disinfectant effective against bacterial spores, bacteria, viruses, TB and fungi (mold).
  • Excellent material compatibility that will not damage most surfaces in cleanroom environments, including stainless steel, acrylic, glass, aluminum and chrome.
  • Does not contain alcohol, bleach, quat or silver.
  • Daily cleaning and disinfecting tasks, such as wiping surfaces and mopping floors.
  • Effective component to monthly cleaning protocols.
  • PeridoxRTU is ideal for use in pharmaceutical, biopharmaceutical and medical device facilities.
  • Sterile PeridoxRTU is ideal for pharmaceutical and life science cleanroom environments, including sterile core areas.
  • Wear appropriate gloves and eye protection. Additional PPE may be required based on engineering controls/environment, method of application, and use protocols.

Order Info

Note: This product (19-033-462) is available in Canada only. As an alternative, see Cat. No. 19-084-205.

Specifications

pH 1.9 to 2.2 (at 20°C)
Sterility Sterile
Content And Storage Store in a cool, dry, well ventilated area away from sources of heat, combustible materials, direct sunlight, and incompatible substances. Keep container tightly closed when not in use. Maintain temperature between 0°C and 40°C. Do not allow product to freeze or become overheated. This may cause increased degradation of the product, which will decrease product effectiveness.
Dilution Ready to use. Do not dilute.
Disposable Consult with the US EPA Guidelines listed in 40 CFR Part 261.3 or the EU Directive 2008/98/EC on waste for the classifications of hazardous waste prior to disposal. Furthermore, consult with your state, local, or provincial waste requirements or guidelines, if applicable, to ensure compliance. Arrange disposal in accordance to the EPA and/or state and local guidelines.
Packaging Double bagged with push/pull cap and foaming trigger
Volume (English) 32 oz.