missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perhydrocyclobuta[c]furan-1,3-dione, 97%, Thermo Scientific™
$270.13 - $270.13
Chemical Identifiers
| CAS | 4462-96-8 |
|---|---|
| Molecular Formula | C6H6O3 |
| Molecular Weight (g/mol) | 126.111 |
| InChI Key | NMNZZIMBGSGRPN-UHFFFAOYSA-N |
| Synonym | 3-oxabicyclo 3.2.0 heptane-2,4-dione, perhydrocyclobuta c furan-1,3-dione, 1,2-cyclobutanedicarboxylic anhydride, cyclobutane-1,2-dicarboxylic anhydride, 1,2-cyclobutanedicarboxylic anhydride, cis-,, acmc-1ahmk, 3-oxabicyclo 3.2.0 heptane-2, cis |
| PubChem CID | 138261 |
| IUPAC Name | 3-oxabicyclo[3.2.0]heptane-2,4-dione |
| SMILES | C1CC2C1C(=O)OC2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
BTB13668DA
|
Thermo Scientific Chemicals
BTB13668DA |
1 g |
Each for $270.13
|
|
|||||
Chemical Identifiers
| 4462-96-8 | |
| 126.111 | |
| 3-oxabicyclo 3.2.0 heptane-2,4-dione, perhydrocyclobuta c furan-1,3-dione, 1,2-cyclobutanedicarboxylic anhydride, cyclobutane-1,2-dicarboxylic anhydride, 1,2-cyclobutanedicarboxylic anhydride, cis-,, acmc-1ahmk, 3-oxabicyclo 3.2.0 heptane-2, cis | |
| 3-oxabicyclo[3.2.0]heptane-2,4-dione |
| C6H6O3 | |
| NMNZZIMBGSGRPN-UHFFFAOYSA-N | |
| 138261 | |
| C1CC2C1C(=O)OC2=O |
Specifications
| 4462-96-8 | |
| Amber Glass Bottle | |
| 1 g | |
| NMNZZIMBGSGRPN-UHFFFAOYSA-N | |
| 3-oxabicyclo[3.2.0]heptane-2,4-dione | |
| 138261 | |
| 97% |
| 95% | |
| C6H6O3 | |
| 3-oxabicyclo 3.2.0 heptane-2,4-dione, perhydrocyclobuta c furan-1,3-dione, 1,2-cyclobutanedicarboxylic anhydride, cyclobutane-1,2-dicarboxylic anhydride, 1,2-cyclobutanedicarboxylic anhydride, cis-,, acmc-1ahmk, 3-oxabicyclo 3.2.0 heptane-2, cis | |
| C1CC2C1C(=O)OC2=O | |
| 126.111 | |
| 126.11 | |
| Perhydrocyclobuta[c]furan-1, 3-dione |