missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctanesulfonamide, 95%

Catalog No. AC459640050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459640050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC459640050 Supplier Thermo Scientific Chemicals Supplier No. 459640050

CAS: 754-91-6 | C8H2F17NO2S | 499.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 754-91-6
Molecular Formula C8H2F17NO2S
Molecular Weight (g/mol) 499.15
InChI Key RRRXPPIDPYTNJG-UHFFFAOYSA-N
Synonym perfluorooctanesulfonamide, perfluoroctylsulfonamide, perfluorooctane sulfonamide, pfosa, heptadecafluorooctanesulphonamide, perfluorooctanesulfonic acid amide, unii-80am718fml, 1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro, deethylsulfluramie, perfluorooctanesulphonamide
PubChem CID 69785
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonamide
SMILES C(C(C(C(C(F)(F)S(=O)(=O)N)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 154.0°C to 155.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (mix of isomers) (GC)
Quantity 5 g
Formula Weight 499.15
Percent Purity 95%
Chemical Name or Material Perfluorooctanesulfonamide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 212-046-

RUO – Research Use Only