missing translation for 'onlineSavingsMsg'
Learn More

Perfluorokerosene (Super-high boiling) (50% in PFK Low boiling) [for Mass Spectrometry], TCI America™

Catalog No. P10615G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10615G Supplier TCI America Supplier No. P10615G
Only null left

Chemical Identifiers

MDL Number MFCD00132835

Specifications

Color Colorless
Quantity 5 g
UN Number 1993
Physical Form Liquid
Chemical Name or Material Perfluorokerosene (Super-high boiling) (50% in PFK Low boiling) [for Mass Spectrometry]