missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluorokerosene (PFK), high boiling, 50% in PFK low boiling, for mass spectrometry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.8900g/mL |
| Boiling Point | <180°C |
| Flash Point | 46°C |
| Packaging | Glass bottle |
| Linear Formula | CnF2n+2 |
| Quantity | 1 mL |
| Specific Gravity | 1.89 |
| Grade | For mass spectrometry |
| Chemical Name or Material | Perfluorokerosene (PFK), high boiling, 50% in PFK low boiling |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. |
RUO – Research Use Only