missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Perfluorohexyl)ethylene 96.0+%, TCI America™
$31.68 - $231.14
Chemical Identifiers
| CAS | 25291-17-2 |
|---|---|
| Molecular Formula | C8H3F13 |
| Molecular Weight (g/mol) | 346.091 |
| MDL Number | MFCD00039249 |
| InChI Key | FYQFWFHDPNXORA-UHFFFAOYSA-N |
| Synonym | perfluorohexyl ethylene, 1h,1h,2h-perfluoro-1-octene, tridecafluorohexyl ethylene, 1-octene, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, unii-j4kbi9s48d, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octene, j4kbi9s48d, perfluorohexylethylene, acmc-209gix, c6f13ch=ch2 |
| PubChem CID | 91384 |
| IUPAC Name | 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene |
| SMILES | C=CC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
Chemical Identifiers
| 25291-17-2 | |
| 346.091 | |
| FYQFWFHDPNXORA-UHFFFAOYSA-N | |
| 91384 | |
| C=CC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
| C8H3F13 | |
| MFCD00039249 | |
| perfluorohexyl ethylene, 1h,1h,2h-perfluoro-1-octene, tridecafluorohexyl ethylene, 1-octene, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, unii-j4kbi9s48d, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octene, j4kbi9s48d, perfluorohexylethylene, acmc-209gix, c6f13ch=ch2 | |
| 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene |
Specifications
| 25291-17-2 | |
| 103°C | |
| MFCD00039249 | |
| 1993 | |
| FYQFWFHDPNXORA-UHFFFAOYSA-N | |
| 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene | |
| 91384 | |
| ≥96.0% (GC) | |
| (Perfluorohexyl)ethylene |
| Colorless | |
| C8H3F13 | |
| 5 g | |
| perfluorohexyl ethylene, 1h,1h,2h-perfluoro-1-octene, tridecafluorohexyl ethylene, 1-octene, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, unii-j4kbi9s48d, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octene, j4kbi9s48d, perfluorohexylethylene, acmc-209gix, c6f13ch=ch2 | |
| C=CC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F | |
| 346.091 | |
| 346.09 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-4067
TSCA : Yes