missing translation for 'onlineSavingsMsg'
Learn More

(Perfluorohexyl)ethylene 96.0+%, TCI America™

Catalog No. P11025G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11025G 5 g
P110225G 25 g
P1102100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P11025G Supplier TCI America Supplier No. P11025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25291-17-2
Molecular Formula C8H3F13
Molecular Weight (g/mol) 346.091
MDL Number MFCD00039249
InChI Key FYQFWFHDPNXORA-UHFFFAOYSA-N
Synonym perfluorohexyl ethylene, 1h,1h,2h-perfluoro-1-octene, tridecafluorohexyl ethylene, 1-octene, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, unii-j4kbi9s48d, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octene, j4kbi9s48d, perfluorohexylethylene, acmc-209gix, c6f13ch=ch2
PubChem CID 91384
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene
SMILES C=CC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 103°C
Quantity 5 g
UN Number 1993
Formula Weight 346.09
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (Perfluorohexyl)ethylene
EINECSNumber (2)-4067
TSCA Yes