missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexanoic Acid Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022501
Change view
Click to view available options
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111022501 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022501 Supplier Merck Emd Millipore Supplier No. 4380925MG

Perfluorohexanoic Acid belongs to the class of perfluorinated carboxylic acids (PFCAs) with a 5-carbon perfluoroalkyl chain, and developed as an alternative to the environmental pollutant, perfluorooctanoic acid (PFOA). The (C-F) bond complicates the degradation of perfluorohexanoic acid.

  • Commonly used as a wetting agent, corrosion inhibitor, lubricant, and foam fire extinguishant

Specifications

CAS 307-24-4
Density 1.759 g/mL (at 20°C (literature))
Molecular Formula C6HF11O2
Linear Formula CF3(CF2)4 COOH
Quantity 25 mg
MDL Number MFCD00198040
Synonym Undecafluorohexanoic acid; Perfluorocaproic acid; Perfluorohexanoic acid
Formula Weight 314.05
Percent Purity ≥98% (GC)
Grade Analytical Standard