missing translation for 'onlineSavingsMsg'
Learn More

(Perfluorocyclohexyl)methyl acrylate, 96%, stab. with 50ppm to 100ppm 4-methoxyphenol

Catalog No. p-7059528
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4479306 5 g
AA4479314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4479306 Supplier Thermo Scientific Chemicals Supplier No. 04479306
Only null left

CAS: 40677-94-9 | C10H5F11O2 | 366.13 g/mol

(Perfluorocyclohexyl)methyl acrylate is used in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40677-94-9
Molecular Formula C10H5F11O2
Molecular Weight (g/mol) 366.13
MDL Number MFCD00080614
InChI Key OZGWOALFBHODRB-UHFFFAOYSA-N
Synonym perfluorocyclohexylmethyl acrylate, perfluorocyclohexyl methyl acrylate, undecafluorocyclohexyl methyl prop-2-enoate, 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl prop-2-enoate, acrylic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexylmethyl ester, 1,2,2,3,3,4,4,5,5,6,6-undecakis fluoranyl cyclohexyl methyl prop-2-enoate, 2-propenoic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl ester
PubChem CID 2776049
IUPAC Name (1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate
SMILES C=CC(=O)OCC1(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(F)F)F

Specifications

Color Colorless
Quantity 5 g
Sensitivity Air sensitive
Solubility Information Sparingly soluble in water.
Formula Weight 366.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material (Perfluorocyclohexyl)methyl acrylate, Stabilized with 100ppm 4-methoxyphenol
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only