missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-tert-butanol, 99%

Catalog No. AAB2244214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2244214 25 g
AAB2244206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2244214 Supplier Thermo Scientific Chemicals Supplier No. B2244214

CAS: 2378-02-1 | C4HF9O | 236.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2378-02-1
MDL Number MFCD00042092

Specifications

Melting Point -17°C
Density 1.693
Boiling Point 44°C to 45°C
Flash Point None
Odor Characteristic
Molecular Formula C7H3ClN2O2S
Linear Formula (CF3)3COH
Refractive Index 1.3 00
Quantity 25 g
UN Number UN2810
Beilstein 1841640
Synonym 2-chloro-6-nitrobenzothiazole, 2-chloro-6-nitrobenzo d thiazole, 2-chloro-6-nitro-benzothiazole, benzothiazole, 2-chloro-6-nitro, 2-chloro-6-nitrobenzo thiazole, 6-nitro-2-chlorobenzothiazole, pubchem21727, 6-nitro-2-chlorbenzothiazol, 2-chloro-6-nitrobenzthiazole, 2-chloro-6-nitro-benzothiazol
InChI Key KUCSJGBXJBQHNI-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1[N+](=O)[O-])SC(=N2)Cl
IUPAC Name 2-chloro-6-nitro-1,3-benzothiazole
Molecular Weight (g/mol) 214.62
PubChem CID 75476
Formula Weight 236.04
Percent Purity 99%
Chemical Name or Material Perfluoro-tert-butanol
Show More Show Less
Hazard Category H315-H319-H332-H335
Hazard Statement GHS H Statement
H331-H315-H319-H335
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 219-157-3
RTECSNumber UB6452700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only