missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-n-octane, 98%, Thermo Scientific Chemicals

Catalog No. p-7023817
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
AAA1774114 25 g
1 options

Catalog No. AAA1774114

Supplier: Thermo Scientific Chemicals A1774114

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorooctane may be used in chemical synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.

Chemical Identifiers

CAS 307-34-6
Molecular Formula C8F18
Molecular Weight (g/mol) 438.059
MDL Number MFCD00042083
InChI Key YVBBRRALBYAZBM-UHFFFAOYSA-N
Synonym perfluorooctane, octadecafluorooctane, n-perfluorooctane, octane, octadecafluoro, eftop ef-l 100, unii-6p60zbk0ql, n-perfluoro octane, perfluoro-n-octane, octadecafluoro-n-octane, 6p60zbk0ql
PubChem CID 9387
ChEBI CHEBI:38826
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-octadecafluorooctane
SMILES C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.73
Boiling Point 99°C to 100°C
Refractive Index 1.282
Quantity 25 g
Beilstein 1717142
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 438.06
Percent Purity 98%
Chemical Name or Material Perfluoro-n-octane
EINECSNumber 206-199-2
RTECSNumber RG9701000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only