Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro(methylcyclohexane), 94%, Thermo Scientific Chemicals

Catalog No. AAA1423314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1423314 25 g
AAA1423322 100 g
AAA1423336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1423314 Supplier Thermo Scientific Chemicals Supplier No. A1423314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Perfluoro(methylcyclohexane) is used as a solvent to determine fluorophilicity of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters. It combines with chloroform and acts as an ingredient for fluorous biphase reactions. Furthermore, it finds application as a heat transfer agent, a dielectric fluid and a perfluorocarbon tracer. It acts as a good solvent for gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(methylcyclohexane) is used as a solvent to determine fluorophilicity of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters. It combines with chloroform and acts as an ingredient for ″fluorous″ biphase reactions. Furthermore, it finds application as a heat transfer agent, a dielectric fluid and a perfluorocarbon tracer. It acts as a good solvent for gases.

Solubility
Miscible with acetone and benzene.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 355-02-2
Molecular Formula C7F14
Molecular Weight (g/mol) 350.06
MDL Number MFCD00001468
InChI Key QIROQPWSJUXOJC-UHFFFAOYSA-N
Synonym perfluoro methylcyclohexane, perfluoromethylcyclohexane, flutec pp2, undecafluoro trifluoromethyl cyclohexane, tetradecafluoromethylcyclohexane, cyclohexane, undecafluoro trifluoromethyl, unii-vj9772yw63, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl cyclohexane, cyclohexane, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl, perfluoro methylcyclohexane , pmch
PubChem CID 9637
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(trifluoromethyl)cyclohexane
SMILES FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)F

Specifications

Melting Point -70°C
Density 1.8
Boiling Point 75°C to 76°C
Odor Odorless
Refractive Index 1.3 00
Quantity 25 g
Beilstein 1915981
Solubility Information Miscible with acetone and benzene.
Formula Weight 350.06
Percent Purity 94%
Chemical Name or Material Perfluoro(methylcyclohexane)
Show More Show Less
EINECSNumber 206-573-5
RTECSNumber GV7610000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only