missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Perfluoro(methylcyclohexane)

Catalog No. 30293725G
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30293725G 25 g
302937100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30293725G Supplier Sigma Aldrich Supplier No. 30293725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Mixtures of chloroform and perfluoro(methylcyclohexane) can be used as solvents for fluorous biphase reactions
  • Gas-phase structure of perfluoro(methylcyclohexane) was investigated.perfluoro(methylcyclohexane) was employed as solvent to investigate fluorophilicity of a series of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters

Specifications

CAS 355-02-2
Density 1.787 g/mL (at 25°C (literature))
Boiling Point 76°C (lit.)
Molecular Formula C7F14
Refractive Index n17/D 1.285 (literature)
Linear Formula C6F11CF3
MDL Number MFCD00001468
Quantity 25 g
Synonym (Trifluoromethyl)undecafluorocyclohexane; Perfluoromethylcyclohexane
Molecular Weight (g/mol) 350.05
Percent Purity 90%
Show More Show Less