missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Perfluoro(methylcyclohexane)
Description
- Mixtures of chloroform and perfluoro(methylcyclohexane) can be used as solvents for fluorous biphase reactions
- Gas-phase structure of perfluoro(methylcyclohexane) was investigated.perfluoro(methylcyclohexane) was employed as solvent to investigate fluorophilicity of a series of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters
Specifications
Specifications
| CAS | 355-02-2 |
| Density | 1.787 g/mL (at 25°C (literature)) |
| Boiling Point | 76°C (lit.) |
| Molecular Formula | C7F14 |
| Refractive Index | n17/D 1.285 (literature) |
| Linear Formula | C6F11CF3 |
| Quantity | 100 g |
| MDL Number | MFCD00001468 |
| Synonym | (Trifluoromethyl)undecafluorocyclohexane; Perfluoromethylcyclohexane |
| Molecular Weight (g/mol) | 350.05 |
| Show More |