missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(2-methyl-2-pentene), 97%

Catalog No. AAA1839406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1839406 5 g
AAA1839414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1839406 Supplier Thermo Scientific Chemicals Supplier No. A1839406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1584-03-8 | C6F12 | 300.047 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1584-03-8
Molecular Formula C6F12
Molecular Weight (g/mol) 300.047
MDL Number MFCD00015724
InChI Key FAEGGADNHFKDQX-UHFFFAOYSA-N
Synonym perfluoro-2-methyl-2-pentene, perfluoro 2-methylpent-2-ene, hexafluoropropene dimer, perfluoro-2-methylpent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-2-ene, perfluoro 2-methyl-2-pentene, nonafluoro-2-trifluoromethyl pent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-4-ene, 2-pentene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl-2-pentene
PubChem CID 74105
IUPAC Name 1,1,1,3,4,4,5,5,5-nonafluoro-2-(trifluoromethyl)pent-2-ene
SMILES C(=C(C(C(F)(F)F)(F)F)F)(C(F)(F)F)C(F)(F)F

Specifications

Density 1.62
Boiling Point 48°C to 54°C
Flash Point −12°C (10°F)
Refractive Index ca. 1.3
Quantity 5 g
UN Number UN1993
Formula Weight 300.05
Percent Purity 97%
Chemical Name or Material Perfluoro(2-methyl-2-pentene)
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 216-436-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only