missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride, 97%

Catalog No. p-7046866
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1661114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1661114 Supplier Thermo Scientific Chemicals Supplier No. L1661114
Only null left

CAS: 34761-47-2 | C15F30O5 | 830.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34761-47-2
Molecular Formula C15F30O5
Molecular Weight (g/mol) 830.11
MDL Number MFCD00054660
InChI Key QDHLRGOEEBWEIB-UHFFFAOYNA-N
Synonym perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride, 3,6,9,12-tetraoxapentadecanoyl fluoride, 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis trifluoromethyl, 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis trifluoromethyl-3,6,9,12-tetraoxapentadecanoyl fluoride, acmc-1csyq, 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis trifluoromethyl-3,6,9,12-tetraoxapentadecan-1-oyl fluoride, perfluoro 2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy propoxy propanoyl fluoride, 2,5,8,11-tetrakis trifluoromethyl-3,6,9,12-tetraoxaheptadecafluoropentadecanoic acid fluoride, 3,6,9,12-tetraoxapentadecanoylfluoride,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis trifluoromethyl
PubChem CID 118188
SMILES FC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Density 1.8
Boiling Point 200°C to 203°C
Flash Point None
Refractive Index 1.3
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
IUPAC Name 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis(trifluoromethyl)-3,6,9,12-tetraoxapentadecanoyl fluoride
Formula Weight 830.12
Percent Purity 97%
Chemical Name or Material Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only