missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-iodooctane, 98%

Catalog No. p-7024936
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2020709 10 g
AAB2020718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2020709 Supplier Thermo Scientific Chemicals Supplier No. B2020709

CAS: 507-63-1 | C8F17I | 545.965 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 507-63-1
Molecular Formula C8F17I
Molecular Weight (g/mol) 545.965
MDL Number MFCD00001064
InChI Key KWXGJTSJUKTDQU-UHFFFAOYSA-N
Synonym perfluorooctyl iodide, heptadecafluoro-1-iodooctane, 1-iodoperfluorooctane, perfluoro-n-octyl iodide, 1-iodoheptadecafluorooctane, perfluoro-1-iodooctane, perfluorooctyliodide, heptadecafluoro-n-octyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodo, octane, heptadecafluoro-1-iodo
PubChem CID 10491
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane
SMILES C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 25°C
Density 2.04
Boiling Point 160°C to 161°C
Refractive Index 1.329
Linear Formula CF3(CF2)7I
Quantity 10 g
Beilstein 1717141
Sensitivity Light sensitive
Formula Weight 545.97
Percent Purity 98%
Chemical Name or Material Perfluoro-1-iodooctane
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-079-5
RTECSNumber RG9700050
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only