missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-iododecane, 97%

Catalog No. AAB2184506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2184506 5 g
AAB2184514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2184506 Supplier Thermo Scientific Chemicals Supplier No. B2184506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 423-62-1 | C10F21I | 645.981 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 423-62-1
Molecular Formula C10F21I
Molecular Weight (g/mol) 645.981
MDL Number MFCD00001065
InChI Key UDWBMXSQHOHKOI-UHFFFAOYSA-N
Synonym perfluorodecyl iodide, 1-iodoperfluorodecane, perfluoro-1-iododecane, henicosafluoro-10-iododecane, heneicosafluorodecyl iodide, heneicosafluoro-1-iododecane, decane, heneicosafluoro-1-iodo, iodoperfluorodecane, henicosafluorodecyl iodide, 1-iodohenicosafluorodecane
PubChem CID 67920
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-10-iododecane
SMILES C(C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 63°C to 67°C
Boiling Point 195°C to 200°C
Flash Point None
Quantity 5 g
Beilstein 1810506
Sensitivity Light sensitive
Formula Weight 645.98
Percent Purity 97%
Chemical Name or Material Perfluoro-1-iododecane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-030-5
RTECSNumber MD8710000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only