missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1,2-dimethylcyclobutane, 97%, remainder 1,3-isomer, Thermo Scientific Chemicals

Catalog No. AAL1675714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1675714 25 g
AAL1675722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1675714 Supplier Thermo Scientific Chemicals Supplier No. L1675714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Perfluoro-1,2-dimethylcyclobutane acts as a ligand for Suzuki coupling. It is involved in the infrared multiple-photon decomposition study of its decomposition products induced by the transverse excited atmospheric pressure pulsed carbon dioxide laser (TEA CO2laser) irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro-1,2-dimethylcyclobutane acts as a ligand for Suzuki coupling. It is involved in the infrared multiple-photon decomposition study of its decomposition products induced by the transverse excited atmospheric pressure pulsed carbon dioxide laser (TEA CO2laser) irradiation.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.strong acids and strong bases.

Chemical Identifiers

CAS 2994-71-0
Molecular Formula C6F12
Molecular Weight (g/mol) 300.05
MDL Number MFCD00013737
InChI Key RBTROQHBNLSUTL-UHFFFAOYNA-N
Synonym Dodecafluorodimethylcyclobutane
PubChem CID 12789874
SMILES FC(F)(F)C1(F)C(F)(F)C(F)(F)C1(F)C(F)(F)F

Specifications

Melting Point -32°C
Density 1.62
Boiling Point 45°C
Refractive Index 1.254
Quantity 25 g
Solubility Information Immiscible with water.
IUPAC Name (3R,4R)-1,1,2,2,3,4-hexafluoro-3,4-bis(trifluoromethyl)cyclobutane
Formula Weight 300.05
Percent Purity 97%
Assay remainder 1,3-isomer
Chemical Name or Material Perfluoro-1,2-dimethylcyclobutane
Show More Show Less
EINECSNumber 221-065-3
TSCA Yes

RUO – Research Use Only