Learn More
Perfex™ Lite-N-Tite Angled Upright Brooms
Supplier:  Perfex™ 2200R
 View more versions of this product
Catalog No. 19042013B
Only null left
Requires : Requires 19-042-036 or 19-042-041 handle.
Description
Description
Provides thorough sweeping with a single pass. Angled polypropylene fibers provide uniform surface contact, even when surfaces are irregular. Polypropylene fibers are fused onto a high-impact resistant polymer block, ensuring long-lasting durability and no absorption of odors, liquids, or bacteria. Available with a flagged end for fine sweeping, or with an unflagged end for coarse sweeping and scrubbing on surfaces like concrete, tile, and asphalt. A patented self-locking connection system prevents handle breakage and keeps broom securely attached. Choice of blue, red, and white.
Specifications
Chemical Identifiers
Angled Upright Brush
A wide range of cleaning requirements with Perfex Lite-N-Tite upright brooms
Adjustable Polymer Handle
Polypropylene Fiber
Specifications
Angled Upright Brush
Flagged end
Adjustable Polymer Handle
Red
A wide range of cleaning requirements with Perfex Lite-N-Tite upright brooms
Polypropylene Fiber
Product Suggestions
Product Suggestions