missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Percoll™ PLUS Centrifugation Media
Description
- Gradients can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor
- Low viscosity of media allows cell isolations on preformed gradients to be completed in only a few minutes using low centrifugal forces (200 to 1000xG)
- Iso-osmotic gradients throughout cover a range of densities up to 1.3g/mL
- Nontoxic, almost chemically inert and does not adhere to membranes
- Adjustable to physiological ionic strength and pH; can be buffered within pH range 5.5 to 10.0 without change in properties
- Silica sol coated with covalently linked silane; 15-30nm dia. bead size
- Low endotoxin levels (max. 2 EU/mL); well-suited for cell separation in clinical research applications
For in vitro research use only.
Specifications
Specifications
| Quantity | 1 L |
| Content And Storage | 4–30°C |
| For Use With (Application) | Cell Isolation |
| Viscosity | 15cP |
| Endotoxin Level | 2 EU/mL |
| Osmolality | < 30 mOsm/kg |
| Color | Colorless to Slightly Yellow |
| pH | 8.9 to 9.9 |
| Product Type | Centrifugation Media |
| Density | 1.135 g/mL |
| Show More |