missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Percoll™ Media
Description
- Composed of colloidal silica coated with polyvinylpyrrolidone (PVP)
- Used for separation of cells, subcellular particles and larger viruses under gentle conditions which preserve viability and morphological integrity of cells
- Low viscosity density gradient medium reduces required centrifugal force
- Nontoxic to cells
- Compatible to ionic strength and pH
- Gradients can either be preformed or created by centrifugation at moderate speeds in an angle-head rotor
- Gradients are iso-osmotic throughout and cover a range of densities up to 1.3g/mL
- Buffered within the pH range 5.5 to 10.0 without any changes in properties
- Conductivity (mS/m): max. 100
- Osmolality (mOsm/kg H2O): max. 25
- Viscosity (cP): max 15
Order Info
Avaialble in easy-to-open, resealable 250mL and 1L bottles
Specifications
Specifications
| Quantity | 6 x 1 L |
| For Use With (Application) | Cell Isolation |
| Viscosity | 15cP |
| Endotoxin Level | <0.12EU/mL |
| Osmolality | < 25 mOsm/kg |
| Color | Colorless to Slightly Yellow |
| pH | 8.5 to 9.5 |
| Product Type | Centrifugation Media |
| Density | 1.135 g/mL |
| Form | Liquid |
| Show More |