missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Percoll™ Centrifugation Media

Catalog No. 45001747
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001747 1 L
45001748 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45001747 Supplier Cytiva Supplier No. 17089101
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For separation of cells, subcellular particles, and larger viruses (down to approx. 70S) under gentle conditions that preserve viability and morphological integrity. Cytiva™ Percoll™ Centrifugation Media Percoll is composed of colloidal silica coated with polyvinylpyrrolidone (PVP).

  • Gradients can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor
  • Low viscosity of media allows cell isolations on preformed gradients to be completed in only a few minutes using low centrifugal forces (200 to 1000xG)
  • Iso-osmotic gradients throughout cover a range of densities up to 1.3g/mL
  • Nontoxic, almost chemically inert and does not adhere to membranes
  • Adjustable to physiological ionic strength and pH; can be buffered within pH range 5.5 to 10.0 without change in properties
  • Percoll is a silica sol with nondialyzable PVP coating; 15-30nm dia. bead size

For in vitro research use only.

Specifications

Quantity 1 L
Content And Storage 4–30°C
For Use With (Application) Cell Isolation
Viscosity 15cP
Osmolality < 25 mOsm/kg
Color Colorless to Slightly Yellow
pH 8.5 to 9.5
Product Type Centrifugation Media
Density 1.135 g/mL
Form Liquid