missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, TraceSELECT™ Ultra, 67 to 72%, Solstice

Catalog No. 60044734
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044734 500 mL Poly Bottle
60044733 250 mL Poly Bottle
60044732 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60044734 Supplier Solstice Supplier No. 12415500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TraceSELECT™ Ultra, for ultratrace analysis, 67-72%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Color Colorless
Melting Point -18°C
Boiling Point 203°C
Density 1.66g/cu.cm
Packaging Poly Bottle
Flash Point 113°C (235.4°F)
Quantity 500 mL
Physical Form Liquid
Linear Formula HClO4
UN Number UN1873
Formula Weight 100.46g/mol
Percent Purity 67 to 72%
Grade TraceSELECT™ Ultra
Chemical Name or Material Perchloric Acid
Show More Show Less
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375
Precautionary Statement H271-H290-H302-H314-H373
DOTInformation 8(5.1)/PGI