missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid (TraceMetal™ Grade), Fisher Chemical™

Catalog No. p-20441
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
HDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A511P500 500 mL HDPE Bottle
A511P212 2.5 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A511P500 Supplier Seastar Chemicals Inc Supplier No. S010201TFIQ03
Only null left
  • Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS.
  • Evaluated for 65 metals at < 1 ppb concentrations.
  • Manufactured, purified, and packaged in a Class1000 clean room.
  • Packaged in non-breakable polypropylene bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA. 45 mm bottle neck size.

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH 0.1
Color Colorless
Melting Point -18°C
Boiling Point 203°C
Density 1.660g/cm³
Packaging HDPE Bottle
Vapor Pressure 6.8mmHg at 25°C
Certificate of Analysis Certificate of Lot Analysis included with each shipment.
Quantity 500 mL
Physical Form Liquid
UN Number 1873
Merck Index 15, 7267
Formula Weight 100.46
Percent Purity 65 to 71% (HClO4, w/w)
Grade TraceMetal
Chemical Name or Material Perchloric Acid
Show More Show Less
ShelfLife 3 years
DOTInformation DOT Class 5.1, : Oxidizer