missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, TraceGrade, 67-71%, Spectrum™ Chemical

Catalog No. 18612188
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612188 2.5 L Poly Bottle
18612187 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612188 Supplier Spectrum Chemical Mfg Cor Supplier No. P13492.5LTPL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 2.5 L
Percent Purity 67 to 71%
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.