missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid Solution, Solstice

Catalog No. 60046339
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046339 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60046339 Supplier Solstice Supplier No. 344081L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Volumetric, 0.01 M HClO4 in acetic acid (0.01N), for titration in non-aqueous liquids

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH Acidic
Color Colorless
Boiling Point 119°C (1013hPa)
Density ∽1.06g/cu.cm at 20°C
Odor Stinging
Packaging Glass bottle
Flash Point 40°C (104°F)
Quantity 1 L
Physical Form Liquid
Linear Formula HClO4
UN Number UN2920
Solubility Information Completely miscible in water
Formula Weight 100.46g/mol
Chemical Name or Material Perchloric acid solution, Volumetric, 0.01 M HClO4 in acetic acid (0.01N), for titration in non-aqueous liquids
Show More Show Less
Hazard Statement P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378
DOTInformation 3(8)/PGII