missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 70.0-72.0%, Solstice

Catalog No. 6001164
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001164 500 mL
6001162 1 L
6001163 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001164 Supplier Solstice Supplier No. 30755500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 70.0-72.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Concentration 70 to 72%
Residue after Ignition ≤0.003% (as SO4)
Flash Point 113°C (235.4°F)
Quantity 500 mL
Linear Formula HClO4
UN Number 1873
Formula Weight 100.46g/mol
Grade ACS/puriss. p.a.
Chemical Name or Material Perchloric Acid, Reag. Ph. Eur. reag. ISO
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375
Precautionary Statement H271-H290-H302-H314-H373
RTECSNumber SC7500000