missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, pure, 0.1M solution in acetic acid

Catalog No. p-3734249
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC348630010 1 L Plastic Bottle
AC348630025 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC348630010 Supplier Thermo Scientific Chemicals Supplier No. 348630010
Only null left

CAS: 7601-90-3 | ClHO4 | 100.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Specifications

Chemical Name or Material Perchloric acid
Color Colorless
Name Note 0.1 M Solution in Acetic Acid
CAS 7601-90-3, 64-19-7
Density 1.0600g/mL
Packaging Plastic Bottle
Specific Gravity 1.06
Flash Point 40°C
Quantity 1 L
Molecular Formula ClHO4
Physical Form Liquid
MDL Number MFCD00011325
UN Number 2920
Fieser 01,796; 02,309; 03,220; 06,453; 09,279; 11,402
Merck Index 15, 7267
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
Solubility Information Solubility in water: insoluble.
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES O[Cl](=O)(=O)=O
IUPAC Name perchloric acid
Molecular Weight (g/mol) 100.45
PubChem CID 24247
ChEBI CHEBI:29221
Formula Weight 100.46
Grade Pure
Show More Show Less

RUO – Research Use Only