missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid (Optima™), Fisher Chemical™

Catalog No. p-20433
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
FEP bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A469250 250 mL FEP bottle
A469500 500 mL FEP bottle
A4691 1 L FEP bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A469250 Supplier Seastar Chemicals Inc Supplier No. S020201TFIF01
Only null left
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH 0.1
Color Colorless
Melting Point -18°C
Boiling Point 203°C
Density 1.660g/cm³
Packaging FEP bottle
Vapor Pressure 6.8mmHg at 25°C
Certificate of Analysis Certificate of Lot Analysis included with each bottle.
Quantity 250 mL
Physical Form Liquid
UN Number 1873
Merck Index 15, 7267
Formula Weight 100.46
Grade OPTIMA
Chemical Name or Material Perchloric Acid
Show More Show Less
DOTInformation DOT Class 5.1, : Oxidizer