missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, ACS, 69.0-72.0%, redistilled

Catalog No. p-7080395
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA87963AP 500 mL
AA87963K4 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA87963AP Supplier Thermo Scientific Chemicals Supplier No. 087963AP
Only null left

CAS: 7601-90-3 | ClHO4 | 100.45 g/mol

Perchloric acid is used as a precursor for ammonium perchlorate which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Melting Point -112°C
Boiling Point 203°C
Density 1.67 g/mL
Flash Point 113°C (235°F)
Assay Percent Range 70%
Quantity 500 mL
Physical Form Liquid
UN Number UN1873
Merck Index 14,7153
Solubility Information Miscible with water.
Formula Weight 100.46
Percent Purity 70%
Grade ACS
Chemical Name or Material Perchloric Acid, redistilled, Redistilled
Show More Show Less
Hazard Category H271-H290-H302-H314-H335-H373
Hazard Statement GHS H Statement
H271-H314
Precautionary Statement P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: PERCHLORIC ACID
EINECSNumber 231-512-4
RTECSNumber SC7500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only