missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 70%, Suprapur™, MilliporeSigma™

Catalog No. M1005171000
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1005171000 1 L
M1005170250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1005171000 Supplier Technidata Supplier No. 1.00517.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Melting Point −18°C
Boiling Point 198.7°C (1013 hPa)
Density 1.68 g/cm3 (at 20°C)
Packaging Glass Bottle
Quantity 1 L
Recommended Storage Store at 5 to 30°C