missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 60%, ACS Reagent, Honeywell™ Fluka™

ACS Reagent, 60%

$459.48 - $459.48

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045918
View Documents
Honeywell-Fluka
311413500ML
500 mL Glass Bottle
Each for $459.48
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7601-90-3
100.45
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
CHEBI:29221
O[Cl](=O)(=O)=O

Specifications

-18°C
203°C
Glass Bottle
7mmHg (25°C)
500 mL
HClO4
UN1873
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
100.46g/mol
ACS Reagent
7601-90-3
1.535g/mL at 25°C
≤0.003%
113°C (235.4°F)
ClHO4
MFCD00011325
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
O[Cl](=O)(=O)=O
100.45
CHEBI:29221
60%
Perchloric Acid
Videos
Safety and Handling

Safety and Handling

P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375

H271-H290-H302-H314-H373

DOTInformation : 8(5.1)/PGI

SDS
missing translation for 'documents'

missing translation for 'documents'