missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, 0.1N in Acetic Acid Standardized Solution

Catalog No. p-4824865
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35608K2 1 L
AA35608K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35608K2 Supplier Thermo Scientific Chemicals Supplier No. 035608K2
Only null left

CAS: 7601-90-3 | ClHO4 | 100.45 g/mol

Perchloric acid, 0.1N in acetic acid standardized solution is used in volumetric analysis. It is useful for its super acid properties and it is also used as an etchant for certain metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Appearance Colorless liquid
Color Colorless
Melting Point 16.6°C
Odor Pungent
Vapor Pressure 16 hPa (17mm Hg) at 20°C
Flash Point 39°C
Quantity 1 L
Physical Form Liquid
UN Number 2920
Solubility Information Miscible with water.
Formula Weight 100.46
Chemical Name or Material Perchloric Acid
Show More Show Less
Hazard Category H226-H290-H314-H335
Precautionary Statement P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation DOT Class: 8; Packing Group: II
EINECSNumber 231-512-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only