missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric acid, 0.1N in Acetic Acid Standardized Solution
Description
Perchloric acid, 0.1N in acetic acid standardized solution is used in volumetric analysis. It is useful for its super acid properties and it is also used as an etchant for certain metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Appearance | Colorless liquid |
| Color | Colorless |
| Melting Point | 16.6°C |
| Odor | Pungent |
| Vapor Pressure | 16 hPa (17mm Hg) at 20°C |
| Flash Point | 39°C |
| Quantity | 1 L |
| UN Number | 2920 |
| Physical Form | Liquid |
| Solubility Information | Miscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H290-H314-H335 |
| Precautionary Statement | P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c |
| DOTInformation | DOT Class: 8; Packing Group: II |
| EINECSNumber | 231-512-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only