missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, 0.043 Normal in N-Propanol, Reagents
Perchloric Acid, 0.043 Normal in N-Propanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CP12040020E7
Specifications
| Perchloric Acid, 0.043 Normal in N-Propanol | |
| 71-23-8,7601-90-3,7732-18-5 | |
| HDPE Winpak | |
| 20 L | |
| Liquid | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| propan-1-ol | |
| Reagent |
| Colorless | |
| 100°C | |
| 1.05 | |
| C3H8O | |
| Chlorine Tetraoxide in 1-Propanol | |
| CCCO | |
| 60.10 |
Chemical Identifiers
| 71-23-8 | |
| 60.10 | |
| Chlorine Tetraoxide in 1-Propanol | |
| CCCO |
| C3H8O | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| propan-1-ol |