missing translation for 'onlineSavingsMsg'
Learn More

Pepsin, 1:3,000, Powder, Spectrum™ Chemical

$480.09 - $43066.65

Chemical Identifiers

CAS 9001-75-6
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18-612-171
View Documents
Spectrum Chemical Mfg Cor
P1571100GM
100 g Poly Bottle
Each for $480.09
Only null left
 
18-612-173
View Documents
Spectrum Chemical Mfg Cor
P1571500GM
500 g Poly Bottle
Each for $1,376.53
Only null left
 
18-612-170
View Documents
Spectrum Chemical Mfg Cor
P157110KG
10 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18-612-172
View Documents
Spectrum Chemical Mfg Cor
P157125KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

9001-75-6

Specifications

9001-75-6
Poly Bottle
Reagent
1
100 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.