missing translation for 'onlineSavingsMsg'
Learn More

Peppermint Oil, Rectified, FCC, 5%, Spectrum™ Chemical

Catalog No. 18605017
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
10 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605017 10 kg Amber Glass Bottle
18605014 25 mL Amber Glass Bottle
18605015 100 mL Amber Glass Bottle
18605016 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605017 Supplier Spectrum Chemical Mfg Cor Supplier No. P159510KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1595, 8006-90-4

Spectrum™ Chemical Peppermint Oil, Rectified, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8006-90-4

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Refractive Index 1.459-1.465
Quantity 10 kg
Specific Gravity 0.896-0.908
Percent Purity 5%
Grade FCC
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Peppermint Oil, Rectified