missing translation for 'onlineSavingsMsg'
Learn More

Peppermint Oil, Rectified, FCC, 5%, Spectrum™ Chemical

P1595, 8006-90-4

Supplier:  Spectrum Chemical Mfg Cor P1595100ML

 View more versions of this product

Catalog No. 18605015


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Peppermint Oil, Rectified, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

8006-90-4
Amber Glass Bottle
100 mL
5%
1
1.459-1.465
0.896-0.908
FCC

Chemical Identifiers

8006-90-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions