missing translation for 'onlineSavingsMsg'
Learn More

Peppermint Oil, NF, 5%, Spectrum™ Chemical

PE105, 8006-90-4

$9797.55 - $147851.03

Chemical Identifiers

CAS 8006-90-4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18605012
View Documents
Spectrum Chemical Mfg Cor
PE10510KGBL
10 kg Steel Pail
Each for $9,797.55
Only null left
 
18605013
View Documents
Spectrum Chemical Mfg Cor
PE105180KGBL
180 kg Steel Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Peppermint Oil, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

8006-90-4

Specifications

8006-90-4
Steel Pail
10 kg
NF
1
1.459-1.465
5%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'