missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Pentyl propionate

Catalog No. 4104461L
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4104461L 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4104461L Supplier Sigma Aldrich Supplier No. 4104461L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Please note that sigma-aldrich provides this product to early discovery researchers as part of a collection of unique chemicals
  • Sigma-aldrich does not collect analytical data for this product
  • Buyer assumes responsibility to confirm product identity and/or purity
  • All sales are final.notwithstanding any contrary provision contained in sigma-aldrich's standard terms and conditions of sale or an agreement between sigma-aldrich and buyer, sigma-aldrich sells this product as-is and makes no representation or warranty whatsoever with respect to this product, including any (a) warranty of merchantability; (b) warranty of fitness for a particular purpose; or (c) warranty against infringement of intellectual property rights of a third party; whether arising by law, course of dealing, course of performance, usage of trade or otherwise

Specifications

CAS 624-54-4
Density 0.873 g/mL (at 25°C (literature))
Boiling Point 169°C (lit.)
Molecular Formula C8H16O2
Refractive Index n20/D 1.4074 (literature)
Linear Formula C2H5CO2(CH2)4 CH3
MDL Number MFCD00048849
Quantity 1 g
Synonym Amyl propionate
Molecular Weight (g/mol) 144.21
Percent Purity ≥99%
Show More Show Less