missing translation for 'onlineSavingsMsg'
Learn More

Pentoxifylline 98.0+%, TCI America™

Catalog No. p-7135378
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20505G 5 g
P205025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20505G Supplier TCI America Supplier No. P20505G
Only null left

Chemical Identifiers

CAS 5-6-6493
Molecular Formula C13H18N4O3
Molecular Weight (g/mol) 278.31
MDL Number MFCD00063379
InChI Key BYPFEZZEUUWMEJ-UHFFFAOYSA-N
Synonym pentoxifylline, trental, oxpentifylline, pentoxifyllin, torental, pentoxyphylline, pentoxil, dimethyloxohexylxanthine, pentoxiphyllium, vazofirin
PubChem CID 4740
ChEBI CHEBI:7986
IUPAC Name 3,7-dimethyl-1-(5-oxohexyl)purine-2,6-dione
SMILES CC(=O)CCCCN1C(=O)C2=C(N=CN2C)N(C1=O)C

Specifications

Melting Point 105°C
Color White
Quantity 5 g
Formula Weight 278.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentoxifylline
RTECSNumber XH2475000
TSCA No