missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentoxifylline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 105°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 278.31 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pentoxifylline |
Safety and Handling
| RTECSNumber | XH2475000 |
| TSCA | No |