missing translation for 'onlineSavingsMsg'
Learn More

Pentasodium Diethylenetriaminepentaacetate (ca. 40% in Water, ca. 1.0mol/L), TCI America™

Catalog No. p-7126981
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D187025G 25 g
D1870500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D187025G Supplier TCI America Supplier No. D187025G
Only null left

Chemical Identifiers

CAS 140-01-2
Molecular Formula C14H18N3Na5O10
Molecular Weight (g/mol) 503.26
MDL Number MFCD00051016
InChI Key LQPLDXQVILYOOL-UHFFFAOYSA-I
Synonym pentasodium dtpa, tetralon b, trilon c, versenex 80, pentasodium pentetate, hamp-ex 80, detarex py, kiresuto p, chelest p, plexene d
PubChem CID 8779
IUPAC Name pentasodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 503.26
Physical Form Liquid
Chemical Name or Material Pentasodium Diethylenetriaminepentaacetate (ca. 40% in Water, ca. 1.0mol/L)
EINECSNumber (2)-1274
TSCA Yes